Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P2RX3
Entrez Gene ID
5024
Gene Name
purinergic receptor P2X 3
Gene Aliases
P2X3
Chromosome Location
Chr.11: 57335943 - 57372399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002559.3 NP_002550.2 3 - 4 510 105
XM_011545069.2 XP_011543371.1 4 - 5 1171 105
XM_011545070.2 XP_011543372.1 4 - 5 477 105
XM_011545071.2 XP_011543373.1 3 - 4 1045 105
XM_011545072.2 XP_011543374.1 4 - 5 1303 105
GenBank mRNA AB016608.1 - 3 - 4 326 105
BC069162.1 - 3 - 4 402 105
BC074792.2 - 3 - 4 357 105
BC074793.2 - 3 - 4 357 105
BC109287.2 - 3 - 4 360 105

View Full Product Details