Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCTD
Entrez Gene ID
1635
Gene Name
dCMP deaminase
Gene Aliases
-
Chromosome Location
Chr.4: 182890091 - 182917477 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012732.1 NP_001012750.1 4 - 5 559 92
NM_001921.2 NP_001912.2 4 - 5 536 92
XM_005262778.3 XP_005262835.1 4 - 5 584 92
XM_005262779.3 XP_005262836.1 5 - 6 675 92
XM_005262780.3 XP_005262837.1 6 - 7 683 92
XM_005262781.4 XP_005262838.1 3 - 4 341 92
XM_005262782.3 XP_005262839.1 3 - 4 338 92
XM_006714115.3 XP_006714178.1 4 - 5 482 92
XM_006714116.3 XP_006714179.1 3 - 4 779 92
XM_011531674.2 XP_011529976.1 5 - 6 1169 92
XM_011531675.2 XP_011529977.1 4 - 5 900 92
XM_011531676.2 XP_011529978.1 5 - 6 908 92
XM_017007816.1 XP_016863305.1 5 - 6 651 92
XM_017007817.1 XP_016863306.1 5 - 6 618 92
XM_017007818.1 XP_016863307.1 6 - 7 683 92
XM_017007819.1 XP_016863308.1 6 - 7 1177 92
XM_017007820.1 XP_016863309.1 6 - 7 1040 92
XM_017007821.1 XP_016863310.1 6 - 7 1173 92
XM_017007822.1 XP_016863311.1 6 - 7 518 92
XM_017007823.1 XP_016863312.1 3 - 4 629 92
XM_017007824.1 XP_016863313.1 3 - 4 256 92
GenBank mRNA AK293135.1 - 3 - 4 315 92
AK313221.1 - 4 - 5 461 92
BC001286.1 - 4 - 5 411 92
BC088357.1 - 4 - 5 450 92
BQ228061.1 - 4 - 5 92
BU188090.1 - 3 - 4 365 92
L12136.1 - 3 - 4 362 92

View Full Product Details