Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCTD
Entrez Gene ID
1635
Gene Name
dCMP deaminase
Gene Aliases
-
Chromosome Location
Chr.4: 182890091 - 182917477 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012732.1 NP_001012750.1 5 - 6 660 76
NM_001921.2 NP_001912.2 5 - 6 637 76
XM_005262778.3 XP_005262835.1 5 - 6 685 76
XM_005262779.3 XP_005262836.1 6 - 7 776 76
XM_005262780.3 XP_005262837.1 7 - 8 784 76
XM_005262781.4 XP_005262838.1 4 - 5 442 76
XM_005262782.3 XP_005262839.1 4 - 5 439 76
XM_006714115.3 XP_006714178.1 5 - 6 583 76
XM_006714116.3 XP_006714179.1 4 - 5 880 76
XM_011531674.2 XP_011529976.1 6 - 7 1270 76
XM_011531675.2 XP_011529977.1 5 - 6 1001 76
XM_011531676.2 XP_011529978.1 6 - 7 1009 76
XM_017007816.1 XP_016863305.1 6 - 7 752 76
XM_017007817.1 XP_016863306.1 6 - 7 719 76
XM_017007818.1 XP_016863307.1 7 - 8 784 76
XM_017007819.1 XP_016863308.1 7 - 8 1278 76
XM_017007820.1 XP_016863309.1 7 - 8 1141 76
XM_017007821.1 XP_016863310.1 7 - 8 1274 76
XM_017007822.1 XP_016863311.1 7 - 8 619 76
XM_017007823.1 XP_016863312.1 4 - 5 730 76
XM_017007824.1 XP_016863313.1 4 - 5 357 76
GenBank mRNA AK293135.1 - 4 - 5 416 76
AK310788.1 - 4 - 5 420 76
AK313221.1 - 5 - 6 562 76
BC001286.1 - 5 - 6 512 76
BC088357.1 - 5 - 6 551 76
BU188090.1 - 4 - 5 466 76
L12136.1 - 4 - 5 463 76

View Full Product Details