Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CCDC7
- Entrez Gene ID
-
79741
- Gene Name
- coiled-coil domain containing 7
- Gene Aliases
-
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
- Chromosome Location
-
Chr.10: 32446082 - 32882864 on Build GRCh38
View Full Product Details