The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002129.3 | NP_002120.1 | 1 - 2 | 178 | 63 |
GenBank mRNA | AU129699.1 | - | 1 - 2 | 175 | 63 |
BC000903.2 | - | 1 - 2 | 70 | 63 | |
BC001063.2 | - | 1 - 2 | 38 | 63 | |
X62534.1 | - | 1 - 2 | 199 | 63 |