Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
KCNE2 | NM_172201.1 | 2 | 579 | NP_751951.1 |
AF071002.1 | 1 | 512 | AAD28086.1 | |
AF302095.1 | 2 | 579 | AAG13416.1 | |
BC093892.1 | AAH93892.1 | |||
BC112087.1 | AAI12088.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv7824n54 | Chr21:34352440 - 34535115 on Build GRCh38 | Gain | KCNE1 LOC105372793 KCNE2 RCAN1 C21orf140 LOC388820 SMIM11A |
dgv4426n100 | Chr21:34339322 - 34534919 on Build GRCh38 | Gain | KCNE1 LOC105372793 KCNE2 RCAN1 C21orf140 LOC388820 SMIM11A |
nsv834088 | Chr21:34241420 - 34379776 on Build GRCh38 | Loss | KCNE2 SMIM11A |
nsv1055429 | Chr21:34299853 - 34516938 on Build GRCh38 | Gain | KCNE1 LOC105372793 KCNE2 RCAN1 C21orf140 LOC388820 SMIM11A |
esv3575386 | Chr21:34353927 - 34534570 on Build GRCh38 | Gain | KCNE1 LOC105372793 KCNE2 RCAN1 C21orf140 LOC388820 SMIM11A |
esv2762109 | Chr21:34350696 - 34531755 on Build GRCh38 | Gain | KCNE1 LOC105372793 KCNE2 RCAN1 C21orf140 LOC388820 SMIM11A |