Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RC3H1
Entrez Gene ID
149041
Gene Name
ring finger and CCCH-type domains 1
Gene Aliases
RNF198, ROQUIN
Chromosome Location
Chr.1: 173931084 - 173993072 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300850.1 NP_001287779.1 17 - 18 3218 71
NM_001300851.1 NP_001287780.1 17 - 18 3191 71
NM_001300852.1 NP_001287781.1 17 - 18 3191 71
NM_172071.3 NP_742068.1 17 - 18 3218 71
XM_005244921.3 XP_005244978.1 17 - 18 3155 71
GenBank mRNA AB095945.1 - 17 - 18 3059 71
AK093501.1 - 8 - 9 1367 71
AK122948.1 - 17 - 18 3191 71
BC136784.1 - 17 - 18 3172 71
BC144408.1 - 17 - 18 3145 71

View Full Product Details