Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RC3H1
Entrez Gene ID
149041
Gene Name
ring finger and CCCH-type domains 1
Gene Aliases
RNF198, ROQUIN
Chromosome Location
Chr.1: 173931084 - 173993072 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300850.1 NP_001287779.1 3 - 4 682 84
NM_001300851.1 NP_001287780.1 3 - 4 682 84
NM_001300852.1 NP_001287781.1 3 - 4 682 84
NM_172071.3 NP_742068.1 3 - 4 682 84
XM_005244921.3 XP_005244978.1 3 - 4 682 84
XM_011509231.2 XP_011507533.1 3 - 4 682 84
GenBank mRNA AB095945.1 - 3 - 4 523 84
AK122948.1 - 3 - 4 682 84
BC136784.1 - 3 - 4 636 84

View Full Product Details