The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142641.1 | NP_001136113.1 | 13 - 14 | 1967 | 67 |
XM_005266177.3 | XP_005266234.1 | 16 - 17 | 2067 | 67 | |
XM_011534808.2 | XP_011533110.1 | 16 - 17 | 2126 | 67 | |
XM_011534810.2 | XP_011533112.1 | 16 - 17 | 2131 | 67 | |
GenBank mRNA | AB046765.2 | - | 13 - 14 | 1738 | 67 |
AF490258.1 | - | 6 - 7 | 833 | 67 |