Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A5
Entrez Gene ID
9121
Gene Name
solute carrier family 16 member 5
Gene Aliases
MCT5, MCT6
Chromosome Location
Chr.17: 75087727 - 75106590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271765.1 NP_001258694.1 1 - 2 166 63
GenBank mRNA AK000416.1 - 1 - 2 166 63

View Full Product Details