Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AOX3P-AOX2P
Entrez Gene ID
107161151
Gene Name
AOX3P-AOX2P readthrough
Gene Aliases
AOX3/AOX3L1
Chromosome Location
Chr.2: 200695723 - 200794915 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_135011.1 18 - 19 2722 119
NR_135012.1 17 - 18 2215 119
GenBank mRNA JX431285.1 - 17 - 18 2021 119
JX431286.1 - 18 - 19 2528 119

View Full Product Details