The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018327.2 | NP_060797.2 | 5 - 6 | 1022 | 107 |
XM_011529279.1 | XP_011527581.1 | 5 - 6 | 790 | 107 | |
XM_011529280.2 | XP_011527582.1 | 2 - 3 | 238 | 107 | |
GenBank mRNA | AK300184.1 | - | 5 - 6 | 828 | 107 |
AL844429.1 | - | 4 - 5 | 107 | ||
BC150644.1 | - | 5 - 6 | 981 | 107 | |
CX870585.1 | - | 4 - 5 | 515 | 107 |