The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142343.1 | NP_001135815.1 | 3 - 4 | 583 | 76 |
NM_001142344.1 | NP_001135816.1 | 2 - 3 | 370 | 76 | |
NM_001142345.1 | NP_001135817.1 | 2 - 3 | 197 | 76 | |
GenBank mRNA | AK291800.1 | - | 2 - 3 | 364 | 76 |
BC106927.2 | - | 3 - 4 | 397 | 76 | |
BC106928.1 | - | 3 - 4 | 397 | 76 | |
BP328503.1 | - | 2 - 3 | 197 | 76 | |
DA466082.1 | - | 2 - 3 | 370 | 76 | |
U79526.1 | - | 2 - 3 | 360 | 76 |