The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015267.3 | NP_056082.2 | 18 - 19 | 3083 | 93 |
XM_011538061.2 | XP_011536363.1 | 18 - 19 | 4593 | 93 | |
XM_011538063.2 | XP_011536365.1 | 17 - 18 | 4458 | 93 | |
XM_011538069.2 | XP_011536371.1 | 18 - 19 | 2932 | 93 | |
XM_011538070.2 | XP_011536372.1 | 18 - 19 | 3842 | 93 | |
XM_017019080.1 | XP_016874569.1 | 19 - 20 | 3925 | 93 | |
XM_017019081.1 | XP_016874570.1 | 19 - 20 | 2988 | 93 | |
GenBank mRNA | AB006631.2 | - | 18 - 19 | 3082 | 93 |
BC151245.1 | - | 18 - 19 | 3093 | 93 |