Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TTTY7
- Entrez Gene ID
-
246122
- Gene Name
- testis-specific transcript, Y-linked 7 (non-protein coding)
- Gene Aliases
-
CLONE795723, LINC00129, NCRNA00129, TTTY7A, TTTY7B, TTY7
- Chromosome Location
-
Chr.Y: 6449468 - 6457906 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001534.2 |
|
1 - 2 |
36 |
100 |
- Gene Symbol
- TTTY7B
- Entrez Gene ID
-
100101120
- Gene Name
- testis-specific transcript, Y-linked 7B (non-protein coding)
- Gene Aliases
-
NCRNA00215
- Chromosome Location
-
Chr.Y: 9706824 - 9715262 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003592.1 |
|
1 - 2 |
36 |
100 |
View Full Product Details