The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079512.3 | NP_001072980.1 | 3 - 4 | 534 | 61 |
NM_001318873.1 | NP_001305802.1 | 3 - 4 | 985 | 61 | |
NR_134901.1 | 4 - 5 | 604 | 61 | ||
NR_134902.1 | 3 - 4 | 534 | 61 | ||
NR_134903.1 | 3 - 4 | 496 | 61 | ||
XM_006720944.3 | XP_006721007.1 | 3 - 4 | 642 | 61 | |
XM_011522654.2 | XP_011520956.1 | 3 - 4 | 642 | 61 | |
XM_017023649.1 | XP_016879138.1 | 3 - 4 | 642 | 61 | |
XM_017023650.1 | XP_016879139.1 | 3 - 4 | 842 | 61 | |
XM_017023651.1 | XP_016879140.1 | 3 - 4 | 270 | 61 | |
GenBank mRNA | AK124106.1 | - | 3 - 4 | 842 | 61 |
AK294418.1 | - | 3 - 4 | 525 | 61 | |
AL833564.1 | - | 1 - 2 | 44 | 61 | |
BC050569.1 | - | 3 - 4 | 309 | 61 | |
BC146882.1 | - | 3 - 4 | 340 | 61 | |
BC146886.1 | - | 3 - 4 | 340 | 61 | |
BC171819.1 | - | 4 - 5 | 410 | 61 | |
CN301583.1 | - | 3 - 4 | 504 | 61 |