The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291661.1 | NP_001278590.1 | 24 - 25 | 3797 | 76 |
XM_011533466.1 | XP_011531768.1 | 24 - 25 | 3794 | 76 | |
XM_011533467.1 | XP_011531769.1 | 24 - 25 | 3761 | 76 | |
XM_011533468.1 | XP_011531770.1 | 24 - 25 | 3794 | 76 | |
XM_011533469.1 | XP_011531771.1 | 24 - 25 | 3794 | 76 | |
XM_011533471.2 | XP_011531773.1 | 24 - 25 | 3794 | 76 | |
XM_017005860.1 | XP_016861349.1 | 24 - 25 | 3794 | 76 | |
XM_017005861.1 | XP_016861350.1 | 23 - 24 | 4477 | 76 | |
XM_017005862.1 | XP_016861351.1 | 3 - 4 | 369 | 76 |