The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291661.1 | NP_001278590.1 | 25 - 26 | 3963 | 123 |
XM_011533466.1 | XP_011531768.1 | 25 - 26 | 3960 | 123 | |
XM_011533467.1 | XP_011531769.1 | 25 - 26 | 3927 | 123 | |
XM_011533468.1 | XP_011531770.1 | 25 - 26 | 3960 | 123 | |
XM_011533469.1 | XP_011531771.1 | 25 - 26 | 3960 | 123 | |
XM_011533471.2 | XP_011531773.1 | 25 - 26 | 3960 | 123 | |
XM_017005860.1 | XP_016861349.1 | 25 - 26 | 3960 | 123 | |
XM_017005861.1 | XP_016861350.1 | 24 - 25 | 4643 | 123 | |
XM_017005862.1 | XP_016861351.1 | 4 - 5 | 535 | 123 | |
GenBank mRNA | U53532.1 | - | 3 - 4 | 372 | 123 |