Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZP1
Entrez Gene ID
22917
Gene Name
zona pellucida glycoprotein 1
Gene Aliases
HEL163, OOMD, OOMD1
Chromosome Location
Chr.11: 60867542 - 60875693 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207341.3 NP_997224.2 10 - 11 1672 62
XM_011544852.2 XP_011543154.1 7 - 8 1188 62
XM_011544853.2 XP_011543155.1 8 - 9 1051 62
GenBank mRNA AI208352.1 - 2 - 3 62
AI766401.1 - 2 - 3 62
BC067899.1 - 6 - 7 1271 62
EU794684.1 - 10 - 11 1672 62

View Full Product Details