Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOLGA3
Entrez Gene ID
2802
Gene Name
golgin A3
Gene Aliases
GCP170, MEA-2
Chromosome Location
Chr.12: 132768909 - 132829131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172557.1 NP_001166028.1 14 - 15 3130 65
NM_005895.3 NP_005886.2 14 - 15 3468 65
XM_005266164.4 XP_005266221.1 14 - 15 3075 65
XM_005266167.4 XP_005266224.1 11 - 12 2500 65
XM_006719736.3 XP_006719799.1 14 - 15 3195 65
XM_006719737.3 XP_006719800.1 14 - 15 3205 65
XM_011534793.2 XP_011533095.1 13 - 14 2922 65
XM_011534794.2 XP_011533096.1 6 - 7 1073 65
GenBank mRNA AB027133.1 - 14 - 15 3190 65
AF485338.1 - 13 - 14 2912 65
BC142658.1 - 14 - 15 3130 65
BC146675.1 - 14 - 15 3130 65
D63997.1 - 14 - 15 3178 65
L06148.1 - 4 - 5 602 65

View Full Product Details