The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022097.3 | NP_071380.1 | 2 - 3 | 566 | 67 |
GenBank mRNA | AF146019.1 | - | 2 - 3 | 571 | 67 |
AK313062.1 | - | 2 - 3 | 422 | 67 |