Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHP2
Entrez Gene ID
63928
Gene Name
calcineurin like EF-hand protein 2
Gene Aliases
-
Chromosome Location
Chr.16: 23754627 - 23758951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022097.3 NP_071380.1 3 - 4 649 73
GenBank mRNA AF146019.1 - 3 - 4 654 73
AK290253.1 - 3 - 4 235 73
AK313062.1 - 3 - 4 505 73

View Full Product Details