The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320707.1 | NP_001307636.1 | 19 - 20 | 2965 | 128 |
NM_001320708.1 | NP_001307637.1 | 19 - 20 | 2808 | 128 | |
NM_001320709.1 | NP_001307638.1 | 20 - 21 | 3061 | 128 | |
NM_006256.3 | NP_006247.1 | 20 - 21 | 3109 | 128 | |
XM_011541772.2 | XP_011540074.1 | 15 - 16 | 3447 | 128 | |
XM_017001782.1 | XP_016857271.1 | 19 - 20 | 2874 | 128 | |
XM_017001783.1 | XP_016857272.1 | 19 - 20 | 2868 | 128 | |
GenBank mRNA | AK298595.1 | - | 19 - 20 | 2647 | 128 |
AK300304.1 | - | 20 - 21 | 2960 | 128 | |
AK301066.1 | - | 15 - 16 | 1810 | 128 | |
BC125199.1 | - | 19 - 20 | 2537 | 128 | |
S75548.1 | - | 20 - 21 | 2681 | 128 | |
U33052.1 | - | 20 - 21 | 2681 | 128 |