Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLTSCR1
Entrez Gene ID
29998
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
-
Chromosome Location
Chr.19: 47608196 - 47703277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015711.3 NP_056526.3 13 - 14 3689 92
XM_005258833.4 XP_005258890.1 13 - 14 3732 92
XM_006723180.3 XP_006723243.1 9 - 10 2652 92
XM_011526882.2 XP_011525184.1 9 - 10 4699 92
XM_011526883.2 XP_011525185.1 12 - 13 3555 92
GenBank mRNA AF182077.1 - 8 - 9 3705 92

View Full Product Details