The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005366.1 | NP_001005366.1 | 19 - 20 | 3653 | 89 |
NM_032590.4 | NP_115979.3 | 20 - 21 | 3557 | 89 | |
XM_005253955.3 | XP_005254012.1 | 20 - 21 | 3767 | 89 | |
XM_005253956.3 | XP_005254013.1 | 20 - 21 | 3364 | 89 | |
XM_005253961.4 | XP_005254018.1 | 9 - 10 | 1852 | 89 | |
XM_011538867.2 | XP_011537169.1 | 20 - 21 | 4819 | 89 | |
XM_011538868.2 | XP_011537170.1 | 20 - 21 | 4815 | 89 | |
XM_011538869.2 | XP_011537171.1 | 20 - 21 | 4817 | 89 | |
XM_011538875.2 | XP_011537177.1 | 9 - 10 | 1861 | 89 | |
GenBank mRNA | AB031230.1 | - | 10 - 11 | 2025 | 89 |
AJ459424.1 | - | 20 - 21 | 3529 | 89 | |
AK027440.1 | - | 10 - 11 | 1946 | 89 | |
AK027692.1 | - | 7 - 8 | 1462 | 89 | |
AK127328.1 | - | 19 - 20 | 3653 | 89 | |
AK299827.1 | - | 9 - 10 | 1871 | 89 | |
AL133572.1 | - | 13 - 14 | 4612 | 89 | |
BC008735.2 | - | 6 - 7 | 1327 | 89 |