The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032590.4 | NP_115979.3 | 1 - 2 | 235 | 115 |
XM_011538867.2 | XP_011537169.1 | 1 - 2 | 1488 | 115 | |
XM_011538868.2 | XP_011537170.1 | 1 - 2 | 1484 | 115 | |
XM_011538869.2 | XP_011537171.1 | 1 - 2 | 1486 | 115 | |
GenBank mRNA | AJ459424.1 | - | 1 - 2 | 207 | 115 |
BC115379.1 | - | 1 - 2 | 174 | 115 | |
BC115380.1 | - | 1 - 2 | 174 | 115 | |
BX648928.1 | - | 1 - 2 | 237 | 116 |