The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024674.5 | NP_078950.1 | 3 - 4 | 534 | 63 |
XM_011542148.1 | XP_011540450.1 | 3 - 4 | 524 | 63 | |
GenBank mRNA | AF521099.1 | - | 3 - 4 | 485 | 63 |
AK022519.1 | - | 3 - 4 | 485 | 63 | |
BC028566.2 | - | 3 - 4 | 524 | 63 | |
CN349325.1 | - | 3 - 4 | 536 | 63 |