Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS7
Entrez Gene ID
6000
Gene Name
regulator of G-protein signaling 7
Gene Aliases
-
Chromosome Location
Chr.1: 240775193 - 241357374 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282775.1 NP_001269704.1 5 - 6 716 89
NM_001282778.1 NP_001269707.1 5 - 6 716 89
NM_002924.5 NP_002915.3 5 - 6 716 89
XM_005273218.3 XP_005273275.1 5 - 6 860 89
XM_006711800.3 XP_006711863.1 4 - 5 651 89
XM_011544246.2 XP_011542548.1 4 - 5 310 89
XM_011544247.2 XP_011542549.1 4 - 5 355 89
XM_017002001.1 XP_016857490.1 4 - 5 809 89
XM_017002002.1 XP_016857491.1 6 - 7 735 89
XM_017002003.1 XP_016857492.1 4 - 5 809 89
XM_017002004.1 XP_016857493.1 4 - 5 809 89
XM_017002005.1 XP_016857494.1 5 - 6 1751 89
XM_017002006.1 XP_016857495.1 6 - 7 781 89
XM_017002007.1 XP_016857496.1 6 - 7 712 89
XM_017002008.1 XP_016857497.1 6 - 7 740 89
XM_017002009.1 XP_016857498.1 4 - 5 640 89
XM_017002010.1 XP_016857499.1 5 - 6 425 89
XM_017002011.1 XP_016857500.1 2 - 3 1822 89
XM_017002012.1 XP_016857501.1 2 - 3 1820 89
XM_017002013.1 XP_016857502.1 2 - 3 1819 89
GenBank mRNA AF090116.1 - 5 - 6 571 89
AF493931.1 - 4 - 5 334 89
AK294234.1 - 5 - 6 550 89
AK294354.1 - 5 - 6 691 89
AK314862.1 - 5 - 6 577 89
AY587875.1 - 4 - 5 334 89
BC022009.1 - 5 - 6 669 89
U32439.1 - 3 - 4 188 89

View Full Product Details