The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042555.2 | NP_001036020.1 | 5 - 6 | 479 | 74 |
NM_001278351.1 | NP_001265280.1 | 6 - 7 | 600 | 74 | |
NM_001278353.1 | NP_001265282.1 | 6 - 7 | 543 | 74 | |
NM_001278354.1 | NP_001265283.1 | 5 - 6 | 532 | 74 | |
NM_001278355.1 | NP_001265284.1 | 5 - 6 | 514 | 74 | |
NM_001278356.1 | NP_001265285.1 | 4 - 5 | 418 | 74 | |
NM_001278357.1 | NP_001265286.1 | 2 - 3 | 279 | 74 | |
NM_006654.4 | NP_006645.3 | 5 - 6 | 486 | 74 | |
XM_017018717.1 | XP_016874206.1 | 6 - 7 | 566 | 74 | |
XM_017018718.1 | XP_016874207.1 | 6 - 7 | 560 | 74 | |
GenBank mRNA | AK312477.1 | - | 4 - 5 | 337 | 74 |
BC021562.1 | - | 2 - 3 | 222 | 74 | |
BP217337.1 | - | 5 - 6 | 447 | 74 | |
BP230333.1 | - | 5 - 6 | 452 | 74 | |
BP280775.1 | - | 5 - 6 | 444 | 74 | |
BP311537.1 | - | 4 - 5 | 385 | 74 | |
BP363404.1 | - | 4 - 5 | 386 | 74 | |
BU166503.1 | - | 6 - 7 | 484 | 74 | |
DA721439.1 | - | 5 - 6 | 461 | 74 | |
DB081843.1 | - | 5 - 6 | 487 | 74 | |
DR003634.1 | - | 6 - 7 | 523 | 74 |