The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281300.1 | NP_001268229.1 | 9 - 10 | 1072 | 77 |
NM_031946.6 | NP_114152.3 | 9 - 10 | 1544 | 77 | |
XM_005249943.2 | XP_005250000.1 | 9 - 10 | 1119 | 77 | |
XM_011515780.1 | XP_011514082.1 | 9 - 10 | 1156 | 77 | |
XM_017011732.1 | XP_016867221.1 | 7 - 8 | 734 | 77 | |
XM_017011733.1 | XP_016867222.1 | 7 - 8 | 734 | 77 | |
XM_017011734.1 | XP_016867223.1 | 9 - 10 | 983 | 77 | |
XM_017011735.1 | XP_016867224.1 | 10 - 11 | 1463 | 77 | |
GenBank mRNA | AF359283.3 | - | 10 - 11 | 1119 | 77 |
AF413079.1 | - | 9 - 10 | 1119 | 77 | |
AK055393.1 | - | 9 - 10 | 1036 | 77 | |
AK096352.1 | - | 4 - 5 | 513 | 77 |