Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGAP3
Entrez Gene ID
116988
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3
Chromosome Location
Chr.7: 151085831 - 151144436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281300.1 NP_001268229.1 8 - 9 971 88
NM_031946.6 NP_114152.3 8 - 9 1443 88
XM_011515780.1 XP_011514082.1 8 - 9 1055 88
XM_017011734.1 XP_016867223.1 8 - 9 882 88
GenBank mRNA AF359283.3 - 9 - 10 1018 88
AF413079.1 - 8 - 9 1018 88
AK055393.1 - 8 - 9 935 88
AK096352.1 - 3 - 4 412 88
BC020426.1 - 7 - 8 799 88

View Full Product Details