The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178229.4 | NP_839943.2 | 26 - 27 | 3358 | 133 |
XM_011509198.2 | XP_011507500.1 | 26 - 27 | 3347 | 133 | |
XM_011509200.2 | XP_011507502.1 | 15 - 16 | 2527 | 133 | |
XM_011509201.2 | XP_011507503.1 | 14 - 15 | 1916 | 133 | |
XM_017000317.1 | XP_016855806.1 | 26 - 27 | 3344 | 133 | |
XM_017000318.1 | XP_016855807.1 | 24 - 25 | 3101 | 133 | |
GenBank mRNA | AB105103.1 | - | 26 - 27 | 3294 | 133 |
AK122653.1 | - | 12 - 13 | 1566 | 133 | |
AY253300.1 | - | 26 - 27 | 3283 | 133 | |
BC131536.1 | - | 26 - 27 | 3316 | 133 |