The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178229.4 | NP_839943.2 | 5 - 6 | 506 | 58 |
XM_011509198.2 | XP_011507500.1 | 5 - 6 | 495 | 58 | |
XM_017000317.1 | XP_016855806.1 | 5 - 6 | 495 | 58 | |
XM_017000318.1 | XP_016855807.1 | 3 - 4 | 249 | 58 | |
GenBank mRNA | AB105103.1 | - | 5 - 6 | 442 | 58 |
AY253300.1 | - | 5 - 6 | 431 | 58 | |
BC131536.1 | - | 5 - 6 | 464 | 58 | |
BG722726.1 | - | 4 - 5 | 424 | 58 |