The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145873.1 | NP_001139345.1 | 4 - 5 | 1080 | 74 |
NM_001768.6 | NP_001759.3 | 1 - 2 | 938 | 74 | |
NM_171827.3 | NP_741969.1 | 1 - 2 | 938 | 74 | |
NR_027353.1 | 2 - 3 | 524 | 74 | ||
GenBank mRNA | AK124156.1 | - | 4 - 5 | 1080 | 74 |
AK300089.1 | - | 2 - 3 | 490 | 74 | |
AY039664.1 | - | 1 - 2 | 228 | 74 | |
BC025715.1 | - | 1 - 2 | 54 | 74 | |
M12824.1 | - | 1 - 2 | 136 | 74 | |
M12828.1 | - | 1 - 2 | 114 | 74 |