Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
Chromosome Location
Chr.8: 86573608 - 86743675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF228520.1 - 12 - 13 1424 82

View Full Product Details