Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
Chromosome Location
Chr.8: 86573608 - 86743675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019098.4 NP_061971.3 7 - 8 952 98
XM_011517138.2 XP_011515440.1 5 - 6 607 98
GenBank mRNA AF228520.1 - 4 - 5 565 98
AF272900.1 - 7 - 8 950 98
BC150601.1 - 7 - 8 930 98

View Full Product Details