The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_120531.1 | 8 - 9 | 1515 | 71 | |
GenBank mRNA | AK096764.1 | - | 1 - 2 | 1858 | 71 |
CD014094.1 | - | 8 - 9 | 1397 | 71 |