Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYAP1
Entrez Gene ID
94056
Gene Name
synapse associated protein 1
Gene Aliases
PRO3113
Chromosome Location
Chr.X: 16719584 - 16762684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032796.3 NP_116185.2 5 - 6 723 81
NR_033181.1 5 - 6 685 81
GenBank mRNA AF168955.1 - 5 - 6 675 81
AF305828.1 - 2 - 3 215 81
AF338728.1 - 5 - 6 675 81
AK027401.1 - 5 - 6 665 81
AK222856.1 - 5 - 6 657 81
AK295322.1 - 5 - 6 660 81
AK303783.1 - 5 - 6 639 81
AK314675.1 - 5 - 6 695 81
BC001468.1 - 5 - 6 654 81
BC014657.1 - 5 - 6 643 81
CR749849.1 - 5 - 6 709 81

View Full Product Details