Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLTSCR1
Entrez Gene ID
29998
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
-
Chromosome Location
Chr.19: 47608196 - 47703277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015711.3 NP_056526.3 5 - 6 341 56
XM_005258833.4 XP_005258890.1 5 - 6 384 56
XM_006723180.3 XP_006723243.1 5 - 6 384 56
XM_011526883.2 XP_011525185.1 5 - 6 384 56
GenBank mRNA BC032065.1 - 5 - 6 341 56
BX362862.2 - 4 - 5 255 56

View Full Product Details