The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004540.3 | NP_004531.2 | 7 - 8 | 1144 | 102 |
XM_011529575.2 | XP_011527877.1 | 8 - 9 | 1619 | 102 | |
XM_011529576.2 | XP_011527878.1 | 8 - 9 | 1196 | 102 | |
XM_011529579.2 | XP_011527881.1 | 7 - 8 | 1120 | 102 | |
XM_011529580.2 | XP_011527882.1 | 8 - 9 | 1619 | 102 | |
XM_011529581.2 | XP_011527883.1 | 8 - 9 | 1619 | 102 | |
XM_011529582.2 | XP_011527884.1 | 8 - 9 | 1618 | 102 | |
XM_011529585.2 | XP_011527887.1 | 4 - 5 | 486 | 102 | |
XM_017028353.1 | XP_016883842.1 | 8 - 9 | 1284 | 102 | |
XM_017028354.1 | XP_016883843.1 | 7 - 8 | 1120 | 102 | |
XM_017028355.1 | XP_016883844.1 | 7 - 8 | 1120 | 102 | |
XM_017028356.1 | XP_016883845.1 | 7 - 8 | 1019 | 102 | |
XM_017028357.1 | XP_016883846.1 | 7 - 8 | 1019 | 102 | |
XM_017028358.1 | XP_016883847.1 | 8 - 9 | 1623 | 102 | |
GenBank mRNA | AK299054.1 | - | 4 - 5 | 657 | 102 |
AK302870.1 | - | 8 - 9 | 1123 | 102 | |
BC052946.1 | - | 7 - 8 | 1144 | 102 | |
U75330.1 | - | 7 - 8 | 964 | 102 |