The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000750.3 | NP_000741.1 | 5 - 6 | 1446 | 68 |
XM_011521186.2 | XP_011519488.1 | 7 - 8 | 1476 | 68 | |
XM_011521187.2 | XP_011519489.1 | 6 - 7 | 1383 | 68 | |
XM_011521190.2 | XP_011519492.1 | 4 - 5 | 1614 | 68 | |
XM_011521191.2 | XP_011519493.1 | 4 - 5 | 1194 | 68 | |
XM_011521192.2 | XP_011519494.1 | 5 - 6 | 1587 | 68 | |
XM_017021885.1 | XP_016877374.1 | 5 - 6 | 1541 | 68 | |
XM_017021886.1 | XP_016877375.1 | 5 - 6 | 3348 | 68 | |
GenBank mRNA | BC096080.1 | - | 5 - 6 | 1449 | 68 |
BC096082.1 | - | 5 - 6 | 1449 | 68 | |
U48861.1 | - | 5 - 6 | 1454 | 68 | |
U62439.1 | - | 5 - 6 | 1420 | 68 | |
X68275.1 | - | 4 - 5 | 1194 | 68 | |
Y08416.1 | - | 5 - 6 | 1425 | 68 |