The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159.3 | NP_001150.3 | 13 - 14 | 1369 | 70 |
XM_011511062.1 | XP_011509364.1 | 13 - 14 | 1425 | 70 | |
XM_017003946.1 | XP_016859435.1 | 13 - 14 | 1425 | 70 | |
XM_017003947.1 | XP_016859436.1 | 13 - 14 | 1425 | 70 | |
GenBank mRNA | AB046692.1 | - | 13 - 14 | 1368 | 70 |
BC117179.1 | - | 13 - 14 | 1322 | 70 | |
BC117181.1 | - | 13 - 14 | 1322 | 70 |