The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000245.3 | NP_000236.2 | 20 - 21 | 4131 | 67 |
NM_001127500.2 | NP_001120972.1 | 20 - 21 | 4185 | 67 | |
NM_001324402.1 | NP_001311331.1 | 19 - 20 | 2948 | 67 | |
XM_006715990.2 | XP_006716053.1 | 20 - 21 | 2870 | 67 | |
XM_011516223.1 | XP_011514525.1 | 21 - 22 | 4160 | 67 | |
GenBank mRNA | AB209898.1 | - | 19 - 20 | 2904 | 67 |
AK296974.1 | - | 20 - 21 | 2848 | 67 | |
AK298552.1 | - | 6 - 7 | 740 | 67 | |
BC027979.1 | - | 1 - 2 | 42 | 67 | |
BC130420.1 | - | 20 - 21 | 4094 | 67 | |
BX648782.1 | - | 8 - 9 | 1158 | 67 | |
J02958.1 | - | 20 - 21 | 4177 | 67 | |
M35074.1 | - | 1 - 2 | 131 | 67 | |
X54559.1 | - | 20 - 21 | 4123 | 67 |