The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130417.2 | NP_001123889.1 | 5 - 6 | 859 | 65 |
NM_033262.4 | NP_150287.1 | 7 - 8 | 3156 | 65 | |
NM_058240.3 | NP_489479.1 | 7 - 8 | 3153 | 65 | |
NM_182932.2 | NP_891977.1 | 6 - 7 | 3144 | 65 | |
NM_182936.2 | NP_891981.1 | 4 - 5 | 755 | 65 | |
NM_183002.2 | NP_892114.1 | 7 - 8 | 3162 | 65 | |
NR_104122.1 | 5 - 6 | 3040 | 65 | ||
XM_006720240.3 | XP_006720303.1 | 6 - 7 | 2008 | 65 | |
XM_017021606.1 | XP_016877095.1 | 7 - 8 | 2534 | 65 | |
XM_017021607.1 | XP_016877096.1 | 7 - 8 | 3024 | 65 | |
XM_017021608.1 | XP_016877097.1 | 6 - 7 | 2770 | 65 | |
XM_017021609.1 | XP_016877098.1 | 6 - 7 | 3006 | 65 | |
XM_017021611.1 | XP_016877100.1 | 5 - 6 | 1132 | 65 | |
GenBank mRNA | AF510501.1 | - | 6 - 7 | 3144 | 65 |
AF510502.1 | - | 7 - 8 | 3162 | 65 | |
AF510503.1 | - | 5 - 6 | 3040 | 65 | |
AJ304852.1 | - | 6 - 7 | 2464 | 65 | |
AJ304853.1 | - | 6 - 7 | 2461 | 65 | |
AJ508602.1 | - | 5 - 6 | 1940 | 65 | |
AJ745101.1 | - | 4 - 5 | 718 | 65 | |
AJ745102.1 | - | 5 - 6 | 822 | 65 | |
AK096553.1 | - | 4 - 5 | 685 | 65 | |
AK122728.1 | - | 5 - 6 | 846 | 65 | |
BC036783.1 | - | 6 - 7 | 2947 | 65 | |
BC142969.1 | - | 5 - 6 | 65 | ||
BC150183.1 | - | 5 - 6 | 2467 | 65 | |
BC150193.1 | - | 5 - 6 | 2467 | 65 |