The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037290.2 | NP_001032367.2 | 10 - 11 | 1515 | 102 |
NM_001256398.1 | NP_001243327.1 | 8 - 9 | 1408 | 102 | |
NM_001256400.1 | NP_001243329.1 | 9 - 10 | 1310 | 102 | |
NM_031938.5 | NP_114144.4 | 10 - 11 | 1627 | 102 | |
GenBank mRNA | AB096252.1 | - | 10 - 11 | 1481 | 102 |
AF276432.1 | - | 10 - 11 | 2124 | 102 | |
AJ290393.1 | - | 10 - 11 | 1477 | 102 | |
AK297853.1 | - | 9 - 10 | 1301 | 102 | |
AK315300.1 | - | 10 - 11 | 1619 | 102 | |
BC041656.1 | - | 8 - 9 | 1371 | 102 |