The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037290.2 | NP_001032367.2 | 2 - 3 | 298 | 89 |
NM_001256397.1 | NP_001243326.1 | 2 - 3 | 298 | 89 | |
NM_001256398.1 | NP_001243327.1 | 2 - 3 | 410 | 89 | |
NM_031938.5 | NP_114144.4 | 2 - 3 | 410 | 89 | |
GenBank mRNA | AB096252.1 | - | 2 - 3 | 264 | 89 |
AF276432.1 | - | 2 - 3 | 907 | 89 | |
AJ290393.1 | - | 2 - 3 | 260 | 89 | |
AK027801.1 | - | 2 - 3 | 293 | 89 | |
AK315300.1 | - | 2 - 3 | 402 | 89 | |
BC041656.1 | - | 2 - 3 | 373 | 89 | |
BX503569.1 | - | 2 - 3 | 298 | 89 |