The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037290.2 | NP_001032367.2 | 1 - 2 | 86 | 123 |
NM_001256397.1 | NP_001243326.1 | 1 - 2 | 86 | 123 | |
GenBank mRNA | AB096252.1 | - | 1 - 2 | 52 | 123 |
AF276432.1 | - | 1 - 2 | 695 | 123 | |
AK027801.1 | - | 1 - 2 | 81 | 123 | |
BX503569.1 | - | 1 - 2 | 86 | 123 |