The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319085.1 | NP_001306014.1 | 3 - 4 | 373 | 59 |
NM_001319087.1 | NP_001306016.1 | 3 - 4 | 373 | 59 | |
NM_001319092.1 | NP_001306021.1 | 3 - 4 | 704 | 59 | |
NM_001319094.1 | NP_001306023.1 | 3 - 4 | 373 | 59 | |
NM_212481.2 | NP_997646.1 | 3 - 4 | 373 | 59 | |
XM_011510499.2 | XP_011508801.1 | 3 - 4 | 2241 | 59 | |
XM_017003183.1 | XP_016858672.1 | 4 - 5 | 719 | 59 | |
GenBank mRNA | AK300173.1 | - | 3 - 4 | 340 | 59 |
AK304566.1 | - | 3 - 4 | 340 | 59 | |
AK307768.1 | - | 3 - 4 | 343 | 59 | |
BC047390.1 | - | 3 - 4 | 707 | 59 | |
BM556343.1 | - | 3 - 4 | 318 | 59 | |
BX396215.2 | - | 3 - 4 | 323 | 59 | |
CR981009.1 | - | 3 - 4 | 328 | 59 | |
M62324.1 | - | 3 - 4 | 325 | 59 |