The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319085.1 | NP_001306014.1 | 5 - 6 | 681 | 47 |
NM_001319087.1 | NP_001306016.1 | 5 - 6 | 678 | 47 | |
NM_001319092.1 | NP_001306021.1 | 5 - 6 | 850 | 47 | |
NM_001319093.1 | NP_001306022.1 | 4 - 5 | 377 | 47 | |
NM_001319096.1 | NP_001306025.1 | 4 - 5 | 380 | 47 | |
NM_212481.2 | NP_997646.1 | 5 - 6 | 519 | 47 | |
XM_011510499.2 | XP_011508801.1 | 5 - 6 | 2387 | 47 | |
XM_017003183.1 | XP_016858672.1 | 6 - 7 | 1024 | 47 | |
XM_017003184.1 | XP_016858673.1 | 4 - 5 | 505 | 47 | |
XM_017003185.1 | XP_016858674.1 | 4 - 5 | 736 | 47 | |
GenBank mRNA | AK300173.1 | - | 5 - 6 | 486 | 47 |
AK304566.1 | - | 4 - 5 | 868 | 47 | |
AK307768.1 | - | 5 - 6 | 489 | 47 | |
BC047390.1 | - | 5 - 6 | 853 | 47 | |
BC067301.1 | - | 4 - 5 | 327 | 47 | |
BQ073595.1 | - | - | 385 | 47 | |
BX396215.2 | - | 5 - 6 | 631 | 47 | |
BX648918.1 | - | 4 - 5 | 348 | 47 | |
CR981009.1 | - | 5 - 6 | 633 | 47 | |
HY076710.1 | - | 4 - 5 | 345 | 47 | |
M62324.1 | - | 5 - 6 | 471 | 47 |