Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYCE1
Entrez Gene ID
93426
Gene Name
synaptonemal complex central element protein 1
Gene Aliases
C10orf94, CT76, POF12, SPGF15
Chromosome Location
Chr.10: 133553900 - 133568291 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143763.1 NP_001137235.1 6 - 7 484 141
NM_001143764.1 NP_001137236.1 6 - 7 484 141
NM_130784.2 NP_570140.1 6 - 7 407 141
GenBank mRNA AK314978.1 - 6 - 7 434 141
AY027807.1 - 6 - 7 407 141
AY027808.1 - 3 - 4 1343 141
BC034821.1 - 6 - 7 434 141
DB053356.1 - 6 - 7 484 141

View Full Product Details